(1R)-(+)-Camphor, 98%
(1R)-(+)-Camphor, 98%
(1R)-(+)-Camphor, 98%
Thermo Scientific Chemicals

(1R)-(+)-Camphor, 98%

CAS: 464-49-3 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10708.22
also known as A10708-22
Price (EUR)
63,30
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
63,30
Each
Chemical Identifiers
CAS464-49-3
IUPAC Name(1R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Molecular FormulaC10H16O
InChI KeyDSSYKIVIOFKYAU-XVKPBYJWSA-N
SMILESCC1(C)[C@H]2CC[C@@]1(C)C(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)174.0-180.0°C
Optical Rotation+41.0 to +45.5° (c=10 in ethanol)
Assay (GC)≥97.5%
Identification (FTIR)Conforms
View more
(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.

Solubility
Soluble in water (0.1 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. G. Grogan.; G. Roberts.; S. Parsons, N. Turner.; S. Flitsch. P450camr, a cytochrome P450 catalysing the stereospecific 6-endo-hydroxylation of (1R)-(+)-camphor. Applied Microbiology and Biotechnology. 2002, 59, (4), 449-454.
  2. V Santhi.; J.Madhusudana Rao. Asymmetric reduction of prochiral ketones using in situ generated oxazaborolidines derived from amino alcohols of (1R)-camphor as catalysts. Tetrahedron: Asymmetry. 2000, 11 (17), 3553-3560.