2,3-Dichlorophenylhydrazine hydrochloride, 97%
2,3-Dichlorophenylhydrazine hydrochloride, 97%
2,3-Dichlorophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals

2,3-Dichlorophenylhydrazine hydrochloride, 97%

CAS: 21938-47-6 | C6H6Cl2N2 | 177.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10717.14
also known as A10717-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS21938-47-6
IUPAC Name(2,3-dichlorophenyl)hydrazine
Molecular FormulaC6H6Cl2N2
InChI KeyHPIALSXRVQRGMK-UHFFFAOYSA-N
SMILESNNC1=CC=CC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only