3-Hydroxy-4-nitrobenzoic acid, 98%
3-Hydroxy-4-nitrobenzoic acid, 98%
3-Hydroxy-4-nitrobenzoic acid, 98%
Thermo Scientific Chemicals

3-Hydroxy-4-nitrobenzoic acid, 98%

CAS: 619-14-7 | C7H5NO5 | 183.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10724.06
also known as A10724-06
Price (EUR)
22,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
22,30
Each
Chemical Identifiers
CAS619-14-7
IUPAC Name3-hydroxy-4-nitrobenzoic acid
Molecular FormulaC7H5NO5
InChI KeyXLDLRRGZWIEEHT-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Melting Point (clear melt)232.0-239.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only