2,3,5-Trimethylphenol, 98+%
2,3,5-Trimethylphenol, 98+%
2,3,5-Trimethylphenol, 98+%
2,3,5-Trimethylphenol, 98+%
Thermo Scientific Chemicals

2,3,5-Trimethylphenol, 98+%

CAS: 697-82-5 | C9H12O | 136.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1000 g
Catalog number A10745.22
also known as A10745-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS697-82-5
IUPAC Name2,3,5-trimethylphenol
Molecular FormulaC9H12O
InChI KeyOGRAOKJKVGDSFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)91.0-98.0°C
Appearance (Color)White to pale cream or pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only