Thermo Scientific Chemicals

Kojic acid, 99%, Thermo Scientific Chemicals

Catalog number: A10760.14
25 g, Each
Thermo Scientific Chemicals

Kojic acid, 99%, Thermo Scientific Chemicals

Catalog number: A10760.14
25 g, Each
Quantity
Catalog number: A10760.14
also known as A10760-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
501-30-4
IUPAC Name
5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
Molecular Formula
C6H6O4
InChI Key
BEJNERDRQOWKJM-UHFFFAOYSA-N
SMILES
OCC1=CC(=O)C(O)=CO1
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.5%
Melting Point (clear melt)
151-158?C
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

Kojic acid may be used on cut fruits to prevent oxidative browning, in seafood to preserve pink and red colors and in cosmetics to lighten skin. It is used to treat skin diseases like melasma as it has antibacterial and antifungal properties. It is used as a chelation agent that inhibits melanin production and also as a tyrosine kinase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, acetone (freely soluble), ethanol (freely soluble), methanol, DMSO, ethyl acetate (slightly soluble), chloroform (slightly soluble), pydidine (slightly soluble) and ether (slightly soluble). Insoluble in benzene.

Notes
Should be stored at room temperature. Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text