Thermo Scientific Chemicals

Salicylamide, 98%, Thermo Scientific Chemicals

Catalog number: A10797.30
250 g, Each
Thermo Scientific Chemicals

Salicylamide, 98%, Thermo Scientific Chemicals

Catalog number: A10797.30
250 g, Each
Quantity
Catalog number: A10797.30
also known as A10797-30
Price (USD)
Price: 34.20
Online price: 29.65
Your price:
Quantity
-

Chemical Identifiers

CAS
65-45-2
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
136.0-144.0?C
Appearance (Color)
White to pale pink to gray
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%

Description

Salicylamide is used in combination with both aspirin and caffeine in the over-the-counter pain remedies. It is used as an analgesic and as an antipyretic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Salicylamide is used in combination with both aspirin and caffeine in the over-the-counter pain remedies. It is used as an analgesic and as an antipyretic.

Solubility
Soluble in water (2.06 mg/ml at 25°C), chloroform, dimethyl sulfoxide, alcohol, and methanol (100 mg/ml).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text