1,10-Diiododecane, 97%
1,10-Diiododecane, 97%
1,10-Diiododecane, 97%
Thermo Scientific Chemicals

1,10-Diiododecane, 97%

CAS: 16355-92-3 | C10H20I2 | 394.079 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10803.09
also known as A10803-09
Price (EUR)
33,60
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
33,60
Each
Chemical Identifiers
CAS16355-92-3
IUPAC Name1,10-diiododecane
Molecular FormulaC10H20I2
InChI KeyCKJCTZAIDVFHCX-UHFFFAOYSA-N
SMILESICCCCCCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
Appearance (Color)White to cream to yellow to pale orange
Melting Point (clear melt)27-32?C
1,10-Diiododecane is used as an alkylating agent in the synthesis of symmetrical and unsymmetrical bis-cryptophanes and as a crosslinking reagent for tetrabutylammonium polygalacturonic acid. it is also used to produce decane at temperature of 20°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Diiododecane is used as an alkylating agent in the synthesis of symmetrical and unsymmetrical bis-cryptophanes and as a crosslinking reagent for tetrabutylammonium polygalacturonic acid. it is also used to produce decane at temperature of 20°C.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
RUO – Research Use Only

General References:

  1. Martina Hirayama.; Walter R. Caseri.; Ulrich W. Suter. Reaction of Long-Chain Iodoalkanes with Gold Surfaces. Journal of Colloid and Interface Science. 1998, 202, (1), 167-172.
  2. John C. Bart.; Dennis G. Peters. Electrochemical reduction of 1,10-dihalodecanes at mercury cathodes in dimethylformamide. Journal of Electroanalytical Chemistry and Interfacial Electrochemistry. 1990, 280 (1), 129-144.