1-(2-Thenoyl)-3,3,3-trifluoroacetone, 99% (dry wt.) may cont. up to ca 2% water
1-(2-Thenoyl)-3,3,3-trifluoroacetone, 99% (dry wt.) may cont. up to ca 2% water
1-(2-Thenoyl)-3,3,3-trifluoroacetone, 99% (dry wt.) may cont. up to ca 2% water
Thermo Scientific Chemicals

1-(2-Thenoyl)-3,3,3-trifluoroacetone, 99% (dry wt.) may cont. up to ca 2% water

CAS: 326-91-0 | C8H5F3O2S | 222.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10808.36
also known as A10808-36
Price (EUR)
876,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
876,00
Each
Chemical Identifiers
CAS326-91-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or cream
FormCrystals or powder or crystalline powder and/or lumps or fused solid
Assay (GC)≥98.5%
CommentMay contain a small quantity of fine red particulate matter from the manufacturing process.
Water Content (Karl Fischer Titration)≤2.5%
1-(2-Thenoyl)-3,3,3-trifluoroacetone is widely used in the analysis of metals such as actinides and lanthanides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Thenoyl)-3,3,3-trifluoroacetone is widely used in the analysis of metals such as actinides and lanthanides.

Solubility
Soluble in methanol and ethanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Chelating ligand. Reagent for Ce, Co, Cr, Cu, Fe, U, V.
  2. Kedari, C. S.; Pandita, S. S.; Parikha, K. J.; Tripathi, S. C.; Gandhi, P. M. Extraction of plutonium (IV) from aqueous nitrate solutions into ligand modified micellar phase (LMMP) of Tergitol 15-S-9 with tri-octylphosphine oxide and separation by ultrafiltration. Desalin. Water Treat. 2014, 52 (1-3), 446-451.
  3. Oliveira, T. C.; Monteiro, R. P. G.; Kastner, G. F.; Barbier, F. B.; Oliveira, A. H. Radiochemical methodologies applied to determination of zirconium isotopes in low-level waste samples from nuclear power plants. J. Radioanal. Nucl. Chem. 2014, 302 (1), 41-47.