1,3-Propanodiol, 99 %, Thermo Scientific Chemicals
1,3-Propanodiol, 99 %, Thermo Scientific Chemicals
1,3-Propanodiol, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Propanodiol, 99 %, Thermo Scientific Chemicals

CAS: 504-63-2 | C3H8O2 | 76.10 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 g
1000 g
5000 g
Número de catálogo A10829.0B
también denominado A10829-0B
Precio (EUR)
154,65
Precio exclusive compra web
172,00
Ahorro 17,35 (10%)
Each
Cantidad:
1000 g
Pedido a granel o personalizado
Precio (EUR)
154,65
Precio exclusive compra web
172,00
Ahorro 17,35 (10%)
Each
Identificadores químicos
CAS504-63-2
IUPAC Namepropane-1,3-diol
Molecular FormulaC3H8O2
InChI KeyYPFDHNVEDLHUCE-UHFFFAOYSA-N
SMILESOCCCO
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.4380-1.4410 @ 20?C
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
1,3-Propanediol is used as a solvent for thin film preparations, in the production of polymers such as polytrimethylene terephthalate, adhesives, laminates, coatings, moldings, aliphatic polyesters, as an antifreeze and in wood paint. It also acts as a reagent for vinyl epoxide synthon, for epoxide ring-opening, for polymerization reactions and for natural product syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El 1,3-propanediol se utiliza como disolvente para preparaciones de película fina, en la producción de polímeros como tereftalato de politrimetileno, adhesivos, laminados, revestimientos, molduras, poliésteres alifáticos, como anticongelante y en pintura de madera. También actúa como reactivo para la sintón de epoxida de vinilo, para la apertura de anillo de epoxida, para reacciones de polimerización y para síntesis de productos naturales.

Solubilidad
Miscible con agua y alcohol.

Notas
Incompatibles con cloruros ácidos, anhídridos ácidos, agentes oxidantes, cloroformatos y agentes reductores.
RUO – Research Use Only

General References:

  1. Used for protection of carbonyl groups as their acetals (1,3-dioxane) derivatives, in the presence of a catalyst, e.g. TsOH: J. Chem. Soc. (C), 244 (1962), pyridine hydrochloride: Bull. Soc. Chim. Fr., 2287 (1972), or Amberlyst™ 15: J. Chem. Soc. Perkin 1, 158 (1979). Comparative studies have indicated that formation of acetals from this diol is slower than for ethylene glycol, as is acid catalyzed hydrolysis of the resulting 1,3-dioxanes of aldehydes in comparison with the 1,3-dioxolanes, whereas for ketones the reverse applies: J. Am. Chem. Soc., 80, 6350 (1958); 90, 1249, 1253 (1968). For a review of the preparation of acetals, see: Synthesis, 501 (1981). Conditions have been described for the protection of acrolein using anhydrous HBr in dichloromethane: Org. Synth. Coll., 7, 59 (1990). For use in the preparation of the cyclic acetal of cyclopropenone, see: Org. Synth. Coll., 8, 173 (1993). The product behaves as a 1,3-dipole in cycloaddition reactions, see Benzyl idenemalononitrile, L02426.
  2. Zhang, J.; Zhao, B.; Pan, Z.; Gu, M.; Punnoose, A. Synthesis of ZnO Nanoparticles with Controlled Shapes, Sizes, Aggregations, and Surface Complex Compounds for Tuning or Switching the Photoluminescence. Cryst. Growth Des. 2015, 15 (7), 3144-3149.
  3. Nakagawa, Y.; Tamura, M.; Tomishige, K. Catalytic materials for the hydrogenolysis of glycerol to 1, 3-propanediol. J. Mater. Chem. A 2014, 2 (19), 6688-6702.