2,2',4'-Trichloroacetophenone, 97%
2,2',4'-Trichloroacetophenone, 97%
2,2',4'-Trichloroacetophenone, 97%
2,2',4'-Trichloroacetophenone, 97%
Thermo Scientific Chemicals

2,2',4'-Trichloroacetophenone, 97%

CAS: 4252-78-2 | C8H5Cl3O | 223.477 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10835.22
also known as A10835-22
Price (EUR)
69,30
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
69,30
Each
Add to cart
Chemical Identifiers
CAS4252-78-2
IUPAC Name2-chloro-1-(2,4-dichlorophenyl)ethan-1-one
Molecular FormulaC8H5Cl3O
InChI KeyVYWPPRLJNVHPEU-UHFFFAOYSA-N
SMILESClCC(=O)C1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)49.0-59.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only