RiboMinus™ Plant Kit for RNA-Seq - FAQs
RiboMinus™ Plant Kit for RNA-Seq - FAQs
View additional product information for RiboMinus™ Plant Kit for RNA-Seq - FAQs (A1083802)
1 product FAQs found
What type of plant samples can be used with the RiboMinus Plant Kit for RNA-Seq?
Plants that can be used with the RiboMinus Plant kit for RNA-Seq include: wheat, maize, rice, barley, soybean, sugar cane, sorghum, potato, cassava, Arabidopsis, tomato, tobacoo, poplar, legume, and moss.