Increase cell survival by more than 50% with new Gibco™ B-27™ Plus Neuronal Culture System.
Looking to improve the performance of your neuronal cultures? Discover the new B-27™ Plus Neuronal Culture System with Neurobasal™ PhysioPlus Medium, designed to support neuronal maintenance and functional activity
Primary Rat Hippocampus Neurons
Gibco™
Primary Rat Hippocampus Neurons
Gibco Primary Rat Hippocampus Neurons are isolated and cryopreserved from day-18 Fisher 344 rat embryos. A flexible, ready-to-use and qualityRead more
Have Questions?
Catalog NumberQuantity
A10841011 x 106 cells
Catalog number A1084101
Price (USD)
-
Quantity:
1 x 106 cells
Gibco Primary Rat Hippocampus Neurons are isolated and cryopreserved from day-18 Fisher 344 rat embryos. A flexible, ready-to-use and quality alternative to fresh neurons, each vial contains 1 million cells per mL. These neurons consistently yield post-thaw viability in the range of 50–80% and have high neuronal purity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Donor AttributesNormal
No. of Cells1 x 106 cells
Product TypeRat Hippocampus Neuron
Quantity1 x 106 cells
Tested ForSterility, Viability, Mycoplasma
AgeFetal
Cell TypeNeural Cells
Culture TypeMammalian Cell Culture
FormCryopreserved
SpeciesRat
TissueHippocampus
Unit SizeEach
Contents & Storage
Store in liquid nitrogen, vapor-phase.
Frequently asked questions (FAQs)
I'm currently culturing neurons. Which media system do you recommend using in conjunction with the Poly-D-Lysine matrix?
We recommend using the B-27 Plus Neuronal Culture System when culturing neurons on Poly-D-Lysine. The B-27 Plus Neuronal Culture System is a complete media system that includes the B-27 Plus Supplement and the Neurobasal Plus Medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What cell types will Poly-D-Lysine support as an attachment matrix?
The Poly-D-Lysine matrix has successfully supported the attachment of primary rodent neurons, iPSC-derived neurons and PC12 cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can primary rat neurons be cryopreserved again after they are cultured?
Primary rat neurons cannot be cryopreserved again because they will not revive.
Can primary rat neurons proliferate?
No, primary rat neurons do not proliferate.
Why does the manual for primary rat neurons have the instruction "Do not expose neurons to air at any time"?
Neurons tend to lift off from the complete medium change, which exposes cells to air. By changing half of the medium, the disturbance can be minimized.