Invitrogen™

Jump-In™TI™ Platform Kit

Catalog number: A10897
Invitrogen™

Jump-In™TI™ Platform Kit

Catalog number: A10897
Catalog Number
A10897
Unit Size
1 kit
Price (USD)
4,475.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A108971 kit
4,475.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
The Jump-In™TI™ Platform Kit enables generation of a Jump-In™TI™ cell line. A Jump-In™TI™ cell line is created by co-transfecting one of three vectors containing the R4 attP target site with the transiently expressed vector for the PhiC31 integrase into your mammalian cell line.
Generating a Jump-In™TI™ cell line is the first step in creating a stable, targeted, and irreversible mammalian cell line expressing your gene of interest. For targeted and irreversible integration of your gene of interest into your Jump-In™TI™ cell line, use the Jump-In™TI™ Gateway™ Vector Kit.
This kit is also a component of the Jump-In™TI™ Gateway™ System.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cloning Method
Gateway™
Delivery Type
Transfection
Expression System
Mammalian
For Use With (Application)
Protein Expression
High-throughput Compatibility
High-throughput Compatible
Key Functions
Stable Cell Line Development, Targeted Integration, Clone your own Promoter
Product Line
Jump-In™
Product Type
Platform Kit
Protein Tag
Untagged
Quantity
1 Kit
Selection Agent (Eukaryotic)
Geneticin™ (G-418), Zeocin™, Blasticidin
Selection Marker Eukaryotic
NeoR, BsdR, ZeoR
Format
Kit
Promoter
None (Promoterless)
Vector
Jump-In Vectors, pJTI

Contents & Storage

The Jump-In™TI™ Platform Kit is composed of three vectors:
pJTI™⁄Bsd Vector (500 ng⁄µl; total: 20 µl),
pJTI™⁄Neo Vector (500 ng⁄µl; total: 20 µl),
pJTI™⁄Zeo Vector (500 ng⁄µl; total 20 µl),
pJTI™ PhiC31 Int Vector (0.5 µg⁄µl; total 20 µl)

Store vectors at -20°C.

Figures