2-Amino-6-fluorobenzoic acid, 98%
2-Amino-6-fluorobenzoic acid, 98%
2-Amino-6-fluorobenzoic acid, 98%
2-Amino-6-fluorobenzoic acid, 98%
Thermo Scientific Chemicals

2-Amino-6-fluorobenzoic acid, 98%

CAS: 434-76-4 | C7H5FNO2 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A10919.03
also known as A10919-03
Price (EUR)
34,90
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
34,90
Each
Add to cart
Chemical Identifiers
CAS434-76-4
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Appearance (Color)White to cream to yellow to pale brown

Used in the synthesis of a novel benzamide with potent neuroleptic activity. 5- fluoro-2-methyl-3,l-benzoxazin-4-one was obtained from 2-amino-6-fluorobenzoic acid. The condensation of 2-amino-6-fluorobenzoic acid methyl ester and (4-fluorophenyl) acetyl chloride with N,N-dimethylamino-4-pyridine in CH 2 Cl 2 to give 2-fluoro-6-[2-(4-fluoro-phenyl)-acetylamino]-benzoic acid methyl ester in 61% yield. 2 Amino 6 fluorobenzoic acid is another easily ac cessible synthon for construction of monofluorinated heterocycles. Reactions of 2,3,4,6-tetra-O-acetyl-n-D_glucopyranosyl bromide (2) with p-hydroxybenzaldehyde, trifluoromethyl and fluoro substi- tuted aniline and 2-amino-6-fluorobenzoic acid gave lO-(Pformyl- pheny1)-2,3,4,6-tetra-O-acetyl-(3-D-glucopyranoside. 2-Fluoro-6-iodobenzoic acid,a loose white crystal,was synthesized by using 2-amino-6-fluorobenzoic acid as the starting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of a novel benzamide with potent neuroleptic activity. 5- fluoro-2-methyl-3,l-benzoxazin-4-one was obtained from 2-amino-6-fluorobenzoic acid. The condensation of 2-amino-6-fluorobenzoic acid methyl ester and (4-fluorophenyl) acetyl chloride with N,N-dimethylamino-4-pyridine in CH 2 Cl 2 to give 2-fluoro-6-[2-(4-fluoro-phenyl)-acetylamino]-benzoic acid methyl ester in 61% yield. 2 Amino 6 fluorobenzoic acid is another easily ac cessible synthon for construction of monofluorinated heterocycles. Reactions of 2,3,4,6-tetra-O-acetyl-n-D_glucopyranosyl bromide (2) with p-hydroxybenzaldehyde, trifluoromethyl and fluoro substi- tuted aniline and 2-amino-6-fluorobenzoic acid gave lO-(Pformyl- pheny1)-2,3,4,6-tetra-O-acetyl-(3-D-glucopyranoside. 2-Fluoro-6-iodobenzoic acid,a loose white crystal,was synthesized by using 2-amino-6-fluorobenzoic acid as the starting material.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Y.S. Shina,; S.J. Song,; S.U. Kanga,; H.S. Hwanga,; J.W. Choid,; B.H. Lee,; Y.-S. Jung,; C.-H. Kim. A novel synthetic compound, 3-amino-3-(4-fluoro-phenyl)-1H-quinoline-2,4-dione, inhibits cisplatin-induced hearing loss by the suppression of reactive oxygen species: In vitro and in vivo study. Neuroscience. 2013, 232 1-12.
  2. Jean-Michel Becht,; Cédric Catala,;Claude Le Drian,; Alain Wagner. Synthesis of Biaryls via Decarboxylative Pd-Catalyzed Cross-Coupling Reaction . Org Letter. 2007, 9 (9), 1781-1783.