Azomethine-H monosodium salt hydrate
Azomethine-H monosodium salt hydrate
Azomethine-H monosodium salt hydrate
Thermo Scientific Chemicals

Azomethine-H monosodium salt hydrate

CAS: 206752-32-1 | C17H12NNaO8S2 | 445.39 g/mol
製品番号(カタログ番号) A10927.06
または、製品番号A10927-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS206752-32-1
IUPAC Namesodium 4-hydroxy-5-{[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-7-sulfonaphthalene-2-sulfonate
Molecular FormulaC17H12NNaO8S2
InChI KeyGRYBZNLNHVSHPY-UHFFFAOYSA-M
SMILES[Na+].OC1=C2C(NC=C3C=CC=CC3=O)=CC(=CC2=CC(=C1)S([O-])(=O)=O)S(O)(=O)=O
さらに表示
Appearance (Color)Orange
Assay (ex Sulphated Ash)> 85.0% (with reference to dried material)
Water Content (Karl Fischer Titration)≤ 8.0%
Formpowder
Azomethine-H monosodium salt hydrate is a reagent used for the colorimetric determination of boron in soil, plants, composts, manure, water and nutrient solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azomethine-H monosodium salt hydrate is a reagent used for the colorimetric determination of boron in soil, plants, composts, manure, water and nutrient solutions.

Solubility
Soluble in water, ethanol and acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
RUO – Research Use Only

General References:

  1. Xin Rui Wang.; Jun Lu.; Dongpeng Yan.; Min Wei.; David G. Evans.; Xue Duan. A photochromic thin film based on salicylideneaniline derivatives intercalated layered double hydroxide. Chemical Physics Letters. 2010, 493, (4-6), 333-339.
  2. Reagent for the colorimetric determination of boron: Analyst, 94, 1135 (1969); Anal. Lett., 8, 559 (1975).