Thermo Scientific Chemicals

Propionyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A10940.0E
2500 g, Each
Thermo Scientific Chemicals

Propionyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A10940.0E
2500 g, Each
Quantity
Catalog number: A10940.0E
also known as A10940-0E
Price (USD)

Chemical Identifiers

CAS
79-03-8
IUPAC Name
propanoyl chloride
Molecular Formula
C3H5ClO
InChI Key
RZWZRACFZGVKFM-UHFFFAOYSA-N
SMILES
CCC(Cl)=O
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4010-1.4070 @ 20?C

Description

Propionyl chloride is used as an intermediate in the production of agrochemicals and pharmaceuticals. It serves as an intermediate for dyes, textile auxiliaries and peroxide compounds. It acts as a crop protecting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propionyl chloride is used as an intermediate in the production of agrochemicals and pharmaceuticals. It serves as an intermediate for dyes, textile auxiliaries and peroxide compounds. It acts as a crop protecting agent.

Notes
Moisture sensitive. Incompatible with alcohols, oxidizing agents, strong bases and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text