1,2-Diiodotetrafluoroethane, 96%, stab. with copper
1,2-Diiodotetrafluoroethane, 96%, stab. with copper
1,2-Diiodotetrafluoroethane, 96%, stab. with copper
Thermo Scientific Chemicals

1,2-Diiodotetrafluoroethane, 96%, stab. with copper

CAS: 354-65-4 | C2F4I2 | 353.825 g/mol
Have Questions?
Catalog NumberQuantity
A10954.06
also known as A10954-06
5 g
Catalog number A10954.06
also known as A10954-06
Price (EUR)
166,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
166,00
Each
Chemical Identifiers
CAS354-65-4
IUPAC Name1,1,2,2-tetrafluoro-1,2-diiodoethane
Molecular FormulaC2F4I2
InChI KeyNZXVPCQHQVWOFD-UHFFFAOYSA-N
SMILESFC(F)(I)C(F)(F)I
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
CommentStabilised with Copper powder
Appearance (Color)Colorless to purple to red to brown to green
Refractive Index1.4920-1.4980 @ 20?C
FormLiquid
1,2-Diiodotetrafluoroethane is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diiodotetrafluoroethane is used as an organic chemical synthesis intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. G.Serboli; B.Minasso. Infra-red study of rotational isomerism in 1,2-diiodotetrafluoroethane. Spectrochimica Acta Part A: Molecular Spectroscopy. 1968, 24,(11), 1813-1819.
  2. J.Liang; D.Drake; M.Roselius. Conformer analysis study of 1,2-diiodotetrafluoroethane by gas chromatography/fourier transform infrared spectroscopy. Applied Spectroscopy. 1994, 48,(2), 206-208.