1,4-Naphthoquinone, 97+% (dry wt.), cont. up to 5% water
1,4-Naphthoquinone, 97+% (dry wt.), cont. up to 5% water
1,4-Naphthoquinone, 97+% (dry wt.), cont. up to 5% water
Thermo Scientific Chemicals

1,4-Naphthoquinone, 97+% (dry wt.), cont. up to 5% water

CAS: 130-15-4 | C10H6O2 | 158.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A10958.22
also known as A10958-22
Price (EUR)
27,70
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
27,70
Each
Chemical Identifiers
CAS130-15-4
IUPAC Name1,4-dihydronaphthalene-1,4-dione
Molecular FormulaC10H6O2
InChI KeyFRASJONUBLZVQX-UHFFFAOYSA-N
SMILESO=C1C=CC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Dark yellow to yellow to brown
Assay (GC)≥97.0%
Water Content (Karl Fischer Titration)≤5%
Identification (FTIR)Conforms
View more
1,4-Naphthoquinone is used as a strong dienophile in Diels-Alder reaction. It is also used as a precursor to anthroquinone and 5-nitro-1,4-naphthalenedione, which finds application in the preparation of aminoanthroquinone and is also used as a dye precursor. It is a basic component of Vitamin K.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Naphthoquinone is used as a strong dienophile in Diels-Alder reaction. It is also used as a precursor to anthroquinone and 5-nitro-1,4-naphthalenedione, which finds application in the preparation of aminoanthroquinone and is also used as a dye precursor. It is a basic component of Vitamin K.

Solubility
Soluble in alkali hydroxide solvents, sparingly soluble in petroleum ether and water. Freely soluble in carbondioxide, acetic acid, benzene and choloroform.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

General References:

  1. Sarkar, S. K.; Sawai, A.; Kanahara, K.; Wentrup, C.; Abe, M.; Gudmundsdottir, A. D. Direct Detection of a Triplet Vinylnitrene, 1,4-Naphthoquinone-2-ylnitrene, in Solution and Cryogenic Matrices. J. Am. Chem. Soc. 2015, 137 (12), 4207-4214.
  2. Sawama, Y.; Kawajiri, T.; Asai, S.; Yasukawa, N.; Shishido, Y.; Monguchi, Y.; Sajiki, H. Biarylmethane and Fused Heterocyclic Arene Synthesis via in Situ Generated o- and/or p-NaphthoquinoneMethides. J. Org. Chem. 2015, 80 (11), 5556-5565.