Thermo Scientific Chemicals

Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals

Catalog number: A10965.18
50 g, Each
Thermo Scientific Chemicals

Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals

Catalog number: A10965.18
50 g, Each
Quantity
Catalog number: A10965.18
also known as A10965-18
Price (USD)
Quantity
-

Chemical Identifiers

CAS
685-87-0
IUPAC Name
1,3-diethyl 2-bromopropanedioate
Molecular Formula
C7H11BrO4
InChI Key
FNJVDWXUKLTFFL-UHFFFAOYSA-N
SMILES
CCOC(=O)C(Br)C(=O)OCC
Assay (unspecified)
≥90.0%

Description

Diethyl bromomalonate is used in perfumes. It is also used to synthesize other compounds such as barbiturates, artificial flavorings, vitamin B1, and vitamin B6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl bromomalonate is used in perfumes. It is also used to synthesize other compounds such as barbiturates, artificial flavorings, vitamin B1, and vitamin B6.

Solubility
Does not mix well with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store at room temperature. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text