3-Iodotoluene, 99%
3-Iodotoluene, 99%
3-Iodotoluene, 99%
Thermo Scientific Chemicals

3-Iodotoluene, 99%

CAS: 625-95-6 | C7H7I | 218.037 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
250 g
Catalog number A10999.30
also known as A10999-30
Price (TWD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS625-95-6
IUPAC Name1-iodo-3-methylbenzene
Molecular FormulaC7H7I
InChI KeyVLCPISYURGTGLP-UHFFFAOYSA-N
SMILESCC1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6035-1.6065 @ 20°C
FormLiquid
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to yellow
3-Iodotoluene was used in the preparartion of indene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodotoluene was used in the preparartion of indene derivatives.

Solubility
Not miscible in water. Soluble in benzene, alcohol and ether.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Zheng-Hui Guan; Zhi-Hui Ren; Lian-Biao Zhao; Yong-Min Liang. Palladium-catalyzed synthesis of indene derivatives via propargylic carbonates with in situ generated organozinc compounds. Organic & Biomolecular Chemistry. 2008, 6,(6), 1040-1045.
  2. O.Buchman; G.Y.Cao; C.T.Peng. Structure assignment by retention index in gas-liquid radiochromatography of substituted cyclohexenes. Journal of Chromatography A. 1984, 312,75-90.