1,4-Dihydroxyanthraquinone, 95%
1,4-Dihydroxyanthraquinone, 95%
1,4-Dihydroxyanthraquinone, 95%
Thermo Scientific Chemicals

1,4-Dihydroxyanthraquinone, 95%

CAS: 81-64-1 | C14H8O4 | 240.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A11010.30
also known as A11010-30
Price (TWD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS81-64-1
IUPAC Name1,4-dihydroxy-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H8O4
InChI KeyGUEIZVNYDFNHJU-UHFFFAOYSA-N
SMILESOC1=CC=C(O)C2=C1C(=O)C1=CC=CC=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥94.0%
Melting Point (clear melt)191.0-203.0?C
Appearance (Color)Orange to red to brown
FormPowder
1,4-Dihydroxyanthraquinone is used to color gasoline and some heating oils. It is used as an intermediate for the synthesis of indanthrene- and alizarin-derived dyes. It is also used to form lake pigments with calcium, barium and lead. It may be used in the synthesis of cyclopentanoids by cyclization of α,β-unsaturated aldehyde with 1, 4 1,4-Dihydroxyanthraquinone. It has been studied as a long range emissive ratiometric fluorescent probe for live cell imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dihydroxyanthraquinone is used to color gasoline and some heating oils. It is used as an intermediate for the synthesis of indanthrene- and alizarin-derived dyes. It is also used to form lake pigments with calcium, barium and lead. It may be used in the synthesis of cyclopentanoids by cyclization of α,β-unsaturated aldehyde with 1, 4 1,4-Dihydroxyanthraquinone. It has been studied as a long range emissive ratiometric fluorescent probe for live cell imaging.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Petronela Pascariu Dorneanu; Mihaela Homocianu; Ionut Radu Tigoianu; Anton Airinei; Mirela Zaltariov; Maria Cazacu. Solvent effects on the photophysical properties of poly[1,4-dihydroxyanthraquinoneimine-1,3-bis(phenylene-ester-methylene)tetramethyldisiloxane]. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2015, 134,218-224.
  2. Makoto Ogawa; Tokuhiko Handa; Kazuyuki Kuroda; Chuzo Kato; Toshiro Tani. Photochemical hole burning of 1,4-dihydroxyanthraquinone intercalated in a pillared layered clay mineral. J. Phys. Chem. 1992, 96,(20), 8116-8119.