Thermo Scientific Chemicals

4-Aminobutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: A11016.0E
2500 g, Each
Thermo Scientific Chemicals

4-Aminobutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: A11016.0E
2500 g, Each
Quantity
Catalog number: A11016.0E
also known as A11016-0E
Price (USD)
Price: 796.00
Online price: 688.65
Your price:
Quantity
-

Chemical Identifiers

CAS
56-12-2
IUPAC Name
4-aminobutanoic acid
Molecular Formula
C4H9NO2
InChI Key
BTCSSZJGUNDROE-UHFFFAOYSA-N
SMILES
NCCCC(O)=O
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Appearance (Color)
White
Form
Powder
Water Content (Karl Fischer Titration)
≤1.5%

Description

In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text