What is the solubility of Sodium butyrate, 98+%?
Sodium butyrate is soluble in water (0.1 g/mL), DMSO (<1 mg/mL), ethanol (~5 mg/mL), and PBS pH7. 2 (~10 mg/mL). Sodium butyrate is insoluble in ether.
Search
View additional product information for Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 568 - FAQs (A11079)
1 product FAQs found