Antimony(III) oxide, 99%
Antimony(III) oxide, 99%
Antimony(III) oxide, 99%
Thermo Scientific Chemicals

Antimony(III) oxide, 99%

CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A11123.36
also known as A11123-36
Price (EUR)
59,30
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
59,30
Each
Chemical Identifiers
CAS1309-64-4
IUPAC Namediantimony(3+) trioxidandiide
Molecular FormulaO3Sb2
InChI KeyGHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Sb+3].[Sb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Supplier's CofA≥98.5 to ≤101.5%
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Jay A. Young.Antimony(III) Oxide. J. Chem. Educ. 2007, 84 (10),1602.
  2. Nelson Belzile,; Yu-Wei Chen1,; Zijian Wang.Oxidation of antimony (III) by amorphous iron and manganese oxyhydroxides. Chemical Geology. 2001, 174 (4),379-387.