Cyclopropanecarbonitrile, 98%
Cyclopropanecarbonitrile, 98%
Cyclopropanecarbonitrile, 98%
Cyclopropanecarbonitrile, 98%
Thermo Scientific Chemicals

Cyclopropanecarbonitrile, 98%

CAS: 5500-21-0 | C4H5N | 67.091 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A11125.18
also known as A11125-18
Price (EUR)
130,00
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
130,00
Each
Add to cart
Chemical Identifiers
CAS5500-21-0
IUPAC Namecyclopropanecarbonitrile
Molecular FormulaC4H5N
InChI KeyAUQDITHEDVOTCU-UHFFFAOYSA-N
SMILESN#CC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4195-1.4235 @ 20?C
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

General References:

  1. Yang, X.; Nath, D.; Fleming, F. F. Chemoselective Alkylations with N- and C-Metalated Nitriles. Org. Lett. 2015, 17 (19), 4906-4909.
  2. Huang, H.; Ji, X.; Xiao, F.; Deng, G. J. Copper(I)/Lewis acid triggered ring-opening coupling reaction of cyclopropenes with nitriles. RSC Adv. 2015, 5 (33), 26335-26338.