Thermo Scientific Chemicals

Cyclopropanecarbonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A11125.30
250 g, Each
Thermo Scientific Chemicals

Cyclopropanecarbonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A11125.30
250 g, Each
Quantity
Catalog number: A11125.30
also known as A11125-30
Price (USD)
Price: 455.00
Online price: 386.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5500-21-0
IUPAC Name
cyclopropanecarbonitrile
Molecular Formula
C4H5N
InChI Key
AUQDITHEDVOTCU-UHFFFAOYSA-N
SMILES
N#CC1CC1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4195-1.4235 @ 20?C

Description

Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text