4-n-Propylcyclohexanone, 99%
4-n-Propylcyclohexanone, 99%
4-n-Propylcyclohexanone, 99%
Thermo Scientific Chemicals

4-n-Propylcyclohexanone, 99%

CAS: 40649-36-3 | C9H16O | 140.226 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11126.14
also known as A11126-14
Price (EUR)
122,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
122,00
Each
Chemical Identifiers
CAS40649-36-3
IUPAC Name4-propylcyclohexan-1-one
Molecular FormulaC9H16O
InChI KeyNQEDLIZOPMNZMC-UHFFFAOYSA-N
SMILESCCCC1CCC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.4515-1.4545 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only