1-Heptyne, 99%
1-Heptyne, 99%
1-Heptyne, 99%
Thermo Scientific Chemicals

1-Heptyne, 99%

CAS: 628-71-7 | C7H12 | 96.173 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11130.14
also known as A11130-14
Price (EUR)
72,70
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
72,70
Each
Chemical Identifiers
CAS628-71-7
IUPAC Namehept-1-yne
Molecular FormulaC7H12
InChI KeyYVXHZKKCZYLQOP-UHFFFAOYSA-N
SMILESCCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4080-1.4110 @ 20?C
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
Assay (GC)≥98.5%
1-Heptyne is used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptyne is used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Argentiere, P.C.L.; Cagnola, E.A.; Quiroga, M.E.; Liprandi, D.A. A palladium tetra-coordinated complex as catalyst in the selective hydrogenation of 1-heptyne. Applied Catalysis A: General. 2002, 226, (1-2), 253-263.
  2. Letchera, T.M.; Moollana, W.C.; Nevinesa, J.A.; Domanska, U. Excess molar volumes and enthalpies of (1-hexyne, or 1-heptyne, or 1-octyne + sulfolane) at the temperature 303.15 K. The Journal of Chemical Thermodynamics. 2000, 32, (5), 579-586.
  3. For triphasic bromination or cyanation in the 1-position, see 1-Octyne, A12440.