1-Bromo-2,2-dimethoxypropane, 98%
1-Bromo-2,2-dimethoxypropane, 98%
1-Bromo-2,2-dimethoxypropane, 98%
1-Bromo-2,2-dimethoxypropane, 98%
Thermo Scientific Chemicals

1-Bromo-2,2-dimethoxypropane, 98%

CAS: 126-38-5 | C5H11BrO2 | 183.045 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A11139.30
also known as A11139-30
Price (EUR)
583,00
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (EUR)
583,00
Each
Add to cart
Chemical Identifiers
CAS126-38-5
IUPAC Name1-bromo-2,2-dimethoxypropane
Molecular FormulaC5H11BrO2
InChI KeySGTITUFGCGGICE-UHFFFAOYSA-N
SMILESCOC(C)(CBr)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.0%
Refractive Index1.4465-1.4510 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only