Thermo Scientific Chemicals

Sodium pyruvate, 99%, Thermo Scientific Chemicals

Catalog number: A11148.30
250 g, Each
Thermo Scientific Chemicals

Sodium pyruvate, 99%, Thermo Scientific Chemicals

Catalog number: A11148.30
250 g, Each
Quantity
Catalog number: A11148.30
also known as A11148-30
Price (USD)
Price: 271.00
Online price: 230.65
Your price:
Quantity
-

Chemical Identifiers

CAS
113-24-6
IUPAC Name
sodium 2-oxopropanoate
Molecular Formula
C3H3NaO3
InChI Key
DAEPDZWVDSPTHF-UHFFFAOYSA-M
SMILES
[Na+].CC(=O)C([O-])=O
Appearance (Color)
White to pale yellow
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Powder or crystalline powder
Loss on Drying
<0.5%

Description

Sodium pyruvate is used in cell culture media as an additional source of energy. It acts as an antioxidant and finds protective effects against oxygen radicals. It serves as an intermediate in many metabolic pathways such as sugar metabolism. It is converted into acetyl coenzyme A and enters the trichloroacetic acid cycle. In addition, it is involved with amino acid metabolism and initiates the Kreb's cycle. It plays an important role as a free radical scavenger.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium pyruvate is used in cell culture media as an additional source of energy. It acts as an antioxidant and finds protective effects against oxygen radicals. It serves as an intermediate in many metabolic pathways such as sugar metabolism. It is converted into acetyl coenzyme A and enters the trichloroacetic acid cycle. In addition, it is involved with amino acid metabolism and initiates the Kreb′s cycle. It plays an important role as a free radical scavenger.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text