3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, 98%
3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, 98%
3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, 98%
Thermo Scientific Chemicals

3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, 98%

CAS: 175136-66-0 | C17H6F12O | 454.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A11164.06
also known as A11164-06
Price (EUR)
214,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
214,00
Each
Chemical Identifiers
CAS175136-66-0
IUPAC Namebis[3,5-bis(trifluoromethyl)phenyl]methanone
Molecular FormulaC17H6F12O
InChI KeyGATWMPGNBWPCIY-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(=CC(=C1)C(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or pale cream
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)136.0-142.0°C
3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, is used as reactive dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′,5,5′-Tetrakis(trifluoromethyl)benzophenone, is used as reactive dye intermediates.

Solubility
Sparingly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Songbai Liu.; Lanny S. Liebeskind. A Simple, Modular Synthesis of Substituted Pyridines. J. Am. Chem. Soc. 2008, 130, (22), 6918-6919.
  2. Zhihui Zhang.; Ying Yu.; Lanny S. Liebeskind. N-Amidation by Copper-Mediated Cross-Coupling of Organostannanes or Boronic Acids with O-Acetyl Hydroxamic Acids. Org. Lett. 2008, 10, (14), 3005-3008.