1-Decyne, 98%
1-Decyne, 98%
1-Decyne, 98%
Thermo Scientific Chemicals

1-Decyne, 98%

CAS: 764-93-2 | C10H18 | 138.254 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11171.06
also known as A11171-06
Price (EUR)
34,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
34,00
Each
Add to cart
Chemical Identifiers
CAS764-93-2
IUPAC Namedec-1-yne
Molecular FormulaC10H18
InChI KeyILLHQJIJCRNRCJ-UHFFFAOYSA-N
SMILESCCCCCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4250-1.4290 @ 20?C
Assay (GC)≥97.5%
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless to pale yellow
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.

Solubility
Not miscible in water.

Notes
Incompatible with strong oxidizing agents. Store at 0-6°C.
RUO – Research Use Only

General References:

  1. Lucy L; Fan; Jiunn- Yann Tang; Alan Wohlman. Investigation of 1-decyne formation in cottonseed oil fried foods. Journal of the American Oil Chemists' Society. 1983, 60 (5), 1115-1119.
  2. For details of hydroalumination (DIBAH) and subsequent palladium(0)-catalyzed coupling with (Z)-1-hexenyl iodide, see: Org. Synth. Coll., 8, 295 (1993).