1-Hexadecanol, 98%
1-Hexadecanol, 98%
1-Hexadecanol, 98%
Thermo Scientific™

1-Hexadecanol, 98%

CAS: 36653-82-4 | C16H34O | 242.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number A11180.0E
also known as A11180-0E
Price (EUR)
99,70
Each
Quantity:
2500 g
Request bulk or custom format
Price (EUR)
99,70
Each
Chemical Identifiers
CAS36653-82-4
IUPAC Namehexadecan-1-ol
Molecular FormulaC16H34O
InChI KeyBXWNKGSJHAJOGX-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormFlakes or pellets
Identification (FTIR)Conforms
Melting Point (clear melt)47.0-53.0?C
Appearance (Color)White
Assay (GC)≥97.5%
1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in alcohol, chloroform, ether

Notes
Keep away from direct sunlight. Incompatible with strong oxidizing agents, strong acids. Store at 2-8°C.
RUO – Research Use Only
Patrick C. Joyce and Mark C. Thies.Vapor-Liquid Equilibria for the Hexane + Hexadecane and Hexane + 1-Hexadecanol Systems at Elevated Temperatures and Pressures. J. Chem. Eng. Data. 1988, 43 (5), 819-822.