Ethyl dodecanoate, 98%
Ethyl dodecanoate, 98%
Ethyl dodecanoate, 98%
Thermo Scientific Chemicals

Ethyl dodecanoate, 98%

CAS: 106-33-2 | C14H28O2 | 228.38 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
產品號碼 A11188.AP
亦稱為 A11188-AP
價格 (TWD)
-
Quantity:
500 mL
Request bulk or custom format
化學識別
CAS106-33-2
IUPAC Nameethyl dodecanoate
Molecular FormulaC14H28O2
InChI KeyMMXKVMNBHPAILY-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(=O)OCC
檢視更多
規格Specification Sheet規格表
Assay (GC)≥97.5%
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Refractive Index1.4290-1.4330 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only