(2-Benzimidazolylthio)acetic acid, 98%
(2-Benzimidazolylthio)acetic acid, 98%
(2-Benzimidazolylthio)acetic acid, 98%
Thermo Scientific Chemicals

(2-Benzimidazolylthio)acetic acid, 98%

CAS: 3042-00-0 | C9H7N2O2S | 207.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11221.14
also known as A11221-14
Price (EUR)
220,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
220,00
Each
Chemical Identifiers
CAS3042-00-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only