4-paracetamol, 98 %, Thermo Scientific Chemicals
4-paracetamol, 98 %, Thermo Scientific Chemicals
4-paracetamol, 98 %, Thermo Scientific Chemicals
4-paracetamol, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-paracetamol, 98 %, Thermo Scientific Chemicals

CAS: 103-90-2 | C8H9NO2 | 151.17 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 g
500 g
1000 g
5000 g
Número de catálogo A11240.0B
también denominado A11240-0B
Precio (USD)
-
Cantidad:
1000 g
Pedido a granel o personalizado
Identificadores químicos
CAS103-90-2
IUPAC NameN-(4-hydroxyphenyl)acetamide
Molecular FormulaC8H9NO2
InChI KeyRZVAJINKPMORJF-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(O)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
FormCrystals or powder or crystalline powder
Melting Point (clear melt)167.0-173.0?C
Appearance (Color)White
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
Cyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.

Solubility
Soluble in alcohol, dimethylformamide, ethylene dichloride, acetone and ethyl acetate. Slightly soluble in water and ether.

Notes
Incompatible with strong oxidizing agents, acids and alkali metals.
RUO – Research Use Only

General References:

  1. Pintal, M.; Kryczka, B.; Marsura, A.; Porwanski, S. Synthesis of bis-cellobiose and bis-glucose derivatives of azacrown macrocycles as hosts in complexes with acetylsalicylic acid and 4-acetamidophenol. Carbohydr. Res. 2014, 386, 18-22.
  2. Albert, A.; Engelhard, C. Chemometric optimization of a low-temperature plasma source design for ambient desorption/ionization mass spectrometry. Spectrochim. Acta, Part B 2015, 105, 109-115.