Thermo Scientific Chemicals

Sodium fumarate, 98%, Thermo Scientific Chemicals

Catalog number: A11276.0E
2500 g, Each
Thermo Scientific Chemicals

Sodium fumarate, 98%, Thermo Scientific Chemicals

Catalog number: A11276.0E
2500 g, Each
Quantity
Catalog number: A11276.0E
also known as A11276-0E
Price (USD)
Price: 470.00
Online price: 399.65
Your price:
Quantity
-

Chemical Identifiers

CAS
17013-01-3
IUPAC Name
disodium (2Z)-but-2-enedioate
Molecular Formula
C4H2Na2O4
InChI Key
MSJMDZAOKORVFC-UAIGNFCESA-L
SMILES
[Na+].[Na+].[O-]C(=O)\C=C/C([O-])=O
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)
≤1.0%
Appearance (Color)
White
Identification (FTIR)
Conforms

Description

Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text