1,1-Diphenylhydrazine hydrochloride, 98%
1,1-Diphenylhydrazine hydrochloride, 98%
1,1-Diphenylhydrazine hydrochloride, 98%
Thermo Scientific Chemicals

1,1-Diphenylhydrazine hydrochloride, 98%

CAS: 530-47-2 | C12H13ClN2 | 220.7 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11289.06
also known as A11289-06
Price (EUR)
31,10
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
31,10
Each
Chemical Identifiers
CAS530-47-2
IUPAC Namehydrogen 1,1-diphenylhydrazine chloride
Molecular FormulaC12H13ClN2
InChI KeyMIVUDWFNUOXEJM-UHFFFAOYSA-N
SMILES[H+].[Cl-].NN(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Freebase GC)≥97.5%
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Appearance (Color)to pale gray to gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only