SeCore® Express B Locus (RUO)
Catalog number: A11306
Invitrogen™  Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
High throughput HLA B Locus typing kit via sequencing based technology for labs handling batches of approximately 300 samples of a single locus.
SeCore® Sequencing Kits directly identify DNA sequence from target HLA genes. Locus specific DNA amplification is accomplished by cycling an amplification mix, FastStart™ Taq DNA Polymerase and sample genomic DNA. The resulting product is treated with ExoSAP-IT™ prior to sequencing, to degrade the unincorporated primers and free nucleotides. The nucleotide chronology and resulting HLA subtype is determined by multicolor, fluorescence-based sequencing.
Contents & storage
Kits contains amplification mix, FastStart™ Taq DNA Polymerase , ExoSAP-IT™, sequencing mixes (including chemistry, primers and enzyme) and precipitation buffer. Store at -20°C in a non-frost free freezer.
Documents
Manuals & protocols