SeCore® Express Cw Locus (RUO)
Catalog number: A11307
Invitrogen™  Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
High throughput HLA Cw Locus typing kit via sequencing based technology for labs handling batches of approximately 300 samples of a single locus.
SeCore® Sequencing Kits directly identify DNA sequence from target HLA genes. Locus specific DNA amplification is accomplished by cycling an amplification mix, FastStart™ Taq DNA Polymerase and sample genomic DNA. The resulting product is treated with ExoSAP-IT™ prior to sequencing, to degrade the unincorporated primers and free nucleotides. The nucleotide chronology and resulting HLA subtype is determined by multicolor, fluorescence-based sequencing.
Contents & storage
Kits contains amplification mix, FastStart™ Taq DNA Polymerase , ExoSAP-IT™, sequencing mixes (including chemistry, primers and enzyme) and precipitation buffer. Store at -20°C in a non-frost free freezer.
Documents
Manuals & protocols