3,4-Dihydroxy-L-phenylalanine, 98+%
3,4-Dihydroxy-L-phenylalanine, 98+%
3,4-Dihydroxy-L-phenylalanine, 98+%
Thermo Scientific Chemicals

3,4-Dihydroxy-L-phenylalanine, 98+%

3-(3,4-Dihydroxyphenyl)-L-alanine (levodopa orL-dopa), CAS # 59-92-7, is a dopamine precursor molecule. | CAS: 59-92-7 | C9H11NO4 | 197.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11311.14
also known as A11311-14
Price (EUR)
156,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
156,00
Each
Chemical Identifiers
CAS59-92-7
IUPAC Name(2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
Molecular FormulaC9H11NO4
InChI KeyWTDRDQBEARUVNC-LURJTMIESA-N
SMILESN[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.0% to ≤102.0% (non-U.S. specification)
Assay from Supplier's CofA≥98.0% (U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
View more
3,4-dihydroxy-L-phenylalanine (DOPA) is used as a cell adhesion molecule in serum-free cultures of anchorage-dependent mammalian cells. It is also used as an anticholinergic drug and used in the treatment of Parkinson's disease. It acts as an intermediate in the biosynthesis of other catecholamines (norepinephrine, epinephrine) and melanin. It acts as a precursor of dopamine product of tyrosine hydroxylase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 3-(3,4-Dihydroxyphenyl)-L-alanine, or L-dopa, is converted to dopamine by DOPA decarboxylase in the body
  • Upon conversion to dopamine, it acts as an excitatory neurotransmitter

Applications

  • L-dopa is useful in the laboratory study of Parkinson's to replenish dopamine in depleted circuits
  • L-dopa has also been used for laboratory research on hypertension and melanocyte activity
RUO – Research Use Only

General References:

  1. Maini Rekdal, V.; Bess, E.N.; Bisanz, J.E.; Turnbaugh, P.J.; Balskus EP. Discovery and inhibition of an interspecies gut bacterial pathway for Levodopa metabolism. Science. 2019, 364(6445), eaau6323.
  2. Hong, C.T.; Chan, L.; Bai, C.H. The Effect of Caffeine on the Risk and Progression of Parkinson's Disease: A Meta-Analysis. Nutrients. 2020, 12(6), 1860.