3,5-Bis(trifluoromethyl)benzeneboronic acid, 97+%
3,5-Bis(trifluoromethyl)benzeneboronic acid, 97+%
3,5-Bis(trifluoromethyl)benzeneboronic acid, 97+%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzeneboronic acid, 97+%

CAS: 73852-19-4 | C8H5BF6O2 | 257.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A11373.03
also known as A11373-03
Price (EUR)
33,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
33,60
Each
Chemical Identifiers
CAS73852-19-4
IUPAC Name[3,5-bis(trifluoromethyl)phenyl]boronic acid
Molecular FormulaC8H5BF6O2
InChI KeyBPTABBGLHGBJQR-UHFFFAOYSA-N
SMILESOB(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.0%
Proton NMRConforms to structure
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Assay (HPLC)≥97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only