Bromobenzene, 99%
Bromobenzene, 99%
Bromobenzene, 99%
Thermo Scientific Chemicals

Bromobenzene, 99%

CAS: 108-86-1 | C6H5Br | 157.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11407.0E
also known as A11407-0E
Price (USD)
154.65
Online Exclusive
172.00
Save 17.35 (10%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
154.65
Online Exclusive
172.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS108-86-1
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤0.5%
Refractive Index1.5580-1.5610 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless to pale yellow or pale blue
FormLiquid
Assay (GC)≥98.5%
View more
Bromobenzene is used as solvent in organic synthesis. It is used to prepare the phenylmagnesium bromide, a Grignard reagent, an intermediate in the preparation of benzoic acid by reacting with carbon dioxide. It is a precursor for making phencyclidine. It is useful for the introduction of phenyl group through palladium-catalyzed coupling reactions viz. the Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromobenzene is used as solvent in organic synthesis. It is used to prepare the phenylmagnesium bromide, a Grignard reagent, an intermediate in the preparation of benzoic acid by reacting with carbon dioxide. It is a precursor for making phencyclidine. It is useful for the introduction of phenyl group through palladium-catalyzed coupling reactions viz. the Suzuki reaction.

Solubility
Miscible with diethyl ether, alcohol, carbon tetrachloride, chloroform and benzene.

Notes
Store in cool place. Avoid inhalation of vapor or mist. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Man, I. C.; Şoriga, Ş. G.; Pârvulescu, V. Theoretical study of the adsorption of bromobenzene and aniline on Cu2O(1 1 0):CuO and Cu2O(1 1 1):CuO surfaces. Chem. Phys. Lett. 2014, 604, 38-45.
  2. Zhang, X.; Borjian, S.; Baird, M. C. An unusual product distribution from the reaction of Pd(η3-1-PhC3H4)(η5-C5H5) with a deficiency of PPh3 in the presence of bromobenzene. Transition Met. Chem. 2015, 40 (2), 125-129.