Thermo Scientific Chemicals

5-Nitro-1,10-phenanthroline, 98%, Thermo Scientific Chemicals

Catalog number: A11428.14
25 g, Each
Thermo Scientific Chemicals

5-Nitro-1,10-phenanthroline, 98%, Thermo Scientific Chemicals

Catalog number: A11428.14
25 g, Each
Quantity
Catalog number: A11428.14
also known as A11428-14
Price (USD)
Price: 1,582.00
Online price: 1,368.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4199-88-6
IUPAC Name
5-nitro-1,10-phenanthroline
Molecular Formula
C12H7N3O2
InChI Key
PDDBTWXLNJNICS-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC2=CC=CN=C2C2=NC=CC=C12
Appearance (Color)
Pale yellow to yellow or cream to pale brown
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)
≤2.5%

Description

5-Nitro-1,10-phenanthroline complexes of iron find wide applications in redox titrations. It is used as a precursor for the synthesis of 5-amino-1,10-phenanthroline, 5-isothiocyanato-1,10-phenanthroline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-1,10-phenanthroline complexes of iron find wide applications in redox titrations. It is used as a precursor for the synthesis of 5-amino-1,10-phenanthroline, 5-isothiocyanato-1,10-phenanthroline.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text