Thermo Scientific Chemicals

2,5-Dihydroxy-1,4-benzoquinone, 98%, Thermo Scientific Chemicals

Catalog number: A11442.14
25 g, Each
Thermo Scientific Chemicals

2,5-Dihydroxy-1,4-benzoquinone, 98%, Thermo Scientific Chemicals

Catalog number: A11442.14
25 g, Each
Quantity
Catalog number: A11442.14
also known as A11442-14
Price (TWD)

Chemical Identifiers

CAS
615-94-1
IUPAC Name
2,5-dihydroxycyclohexa-2,5-diene-1,4-dione
Molecular Formula
C6H4O4
InChI Key
QFSYADJLNBHAKO-UHFFFAOYSA-N
SMILES
OC1=CC(=O)C(O)=CC1=O
Identification (FTIR)
Conforms
Form
Powder
Appearance (Color)
Cream to yellow-orange to brown
Assay (HPLC)
≥97.5%

Description

2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text