Thermo Scientific Chemicals

D-(-)-Penicillamine, 98%, Thermo Scientific Chemicals

Catalog number: A11446.22
100 g, Each
Thermo Scientific Chemicals

D-(-)-Penicillamine, 98%, Thermo Scientific Chemicals

Catalog number: A11446.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11446.22
also known as A11446-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
52-67-5
IUPAC Name
(2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
Molecular Formula
C5H11NO2S
InChI Key
VVNCNSJFMMFHPL-VKHMYHEASA-N
SMILES
CC(C)(S)[C@@H](N)C(O)=O
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White to pale cream
Optical Rotation
-62.5? ? 2? (c=5%, 1N NaOH)

Description

It is used mainly as chelating agent in heavy metal poisoning as in lead, mercury and copper poisoning. It is used in Wilson's disease. It  is used in pharmacy as chelating agent and as immunosuppressant for treating rheumatoid arthritis by reducing the number of T cells, inhibiting the macrophages, by reducing the activity of Interleukin 1 and rheumatoid factor and in preventing crosslinking of the collagen. It acts as an exogenous NOS modulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
It is freely soluble in water, slightly soluble in alcohol, insoluble in chloroform, ether, acetone, benzene, and in carbon tetrachloride.

Notes
Stable under normal conditions. Incompatabile to strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text