Octafluorotoluene, 98+%
Octafluorotoluene, 98+%
Octafluorotoluene, 98+%
Thermo Scientific Chemicals

Octafluorotoluene, 98+%

CAS: 434-64-0 | C7F8 | 236.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11453.06
also known as A11453-06
Price (EUR)
37,10
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
37,10
Each
Chemical Identifiers
CAS434-64-0
IUPAC Name1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
Molecular FormulaC7F8
InChI KeyUSPWUOFNOTUBAD-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.3655-1.3695 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only