Thermo Scientific Chemicals

2-Furoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11454.36
500 g, Each
Thermo Scientific Chemicals

2-Furoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11454.36
500 g, Each
Quantity
Catalog number: A11454.36
also known as A11454-36
Price (USD)
Price: 122.00
Online price: 103.65
Your price:
Quantity
-

Chemical Identifiers

CAS
88-14-2
IUPAC Name
furan-2-carboxylic acid
Molecular Formula
C5H4O3
InChI Key
SMNDYUVBFMFKNZ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CO1
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Appearance (Color)
White to cream to pale brown
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text