Thermo Scientific Chemicals

L-Phenylalanine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A11455.18
50 g, Each
Thermo Scientific Chemicals

L-Phenylalanine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A11455.18
50 g, Each
Quantity
Catalog number: A11455.18
also known as A11455-18
Price (USD)
Price: 202.00
Online price: 171.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7524-50-7
IUPAC Name
hydrogen methyl 2-amino-3-phenylpropanoate chloride
Molecular Formula
C10H14ClNO2
InChI Key
SWVMLNPDTIFDDY-UHFFFAOYNA-N
SMILES
[H+].[Cl-].COC(=O)C(N)CC1=CC=CC=C1
Melting Point (clear melt)
151-162?C
Appearance (Color)
White to pale cream
Assay (Argentometric Titration)
≥97.5 to ≤102.5%
Optical Rotation
+37? to +39? (c=2 in Ethanol)
Identification (FTIR)
Conforms

Description

Methyl L-phenylalaninate hydrochloride is used in pharmaceutical, food,animal feed,cosmetic,bio-chemical research etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl L-phenylalaninate hydrochloride is used in pharmaceutical, food,animal feed,cosmetic,bio-chemical research etc.

Solubility
It is soluble in methanol. (5mg/ ml-clear colorless solution)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents. Stable under recommended storage conditions. Hygroscopic.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text