Bis(cyclopentadienyl)titanium dichloride, 97%
Bis(cyclopentadienyl)titanium dichloride, 97%
Bis(cyclopentadienyl)titanium dichloride, 97%
Thermo Scientific Chemicals

Bis(cyclopentadienyl)titanium dichloride, 97%

CAS: 1271-19-8 | C10H10Cl2Ti-2 | 248.957 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A11456.18
also known as A11456-18
Price (EUR)
131,65
Online Exclusive
146,00
Save 14,35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
131,65
Online Exclusive
146,00
Save 14,35 (10%)
Each
Chemical Identifiers
CAS1271-19-8
IUPAC Nametitanium(2+) bis(cyclopenta-2,4-dien-1-ide) dichloride
Molecular FormulaC10H10Cl2Ti
InChI KeyMKNXBRLZBFVUPV-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ti++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Appearance (Color)Dark red
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
One-pot synthesis of α-methylene-γ-butyrolactones from benzaldehydes and bromomethylacrylates. Bis-(cyclopentadienyl)-titanium dichloride alkylaluminum complexls acts as catalysts for the polymerization of ethylene. Polyethylene prepared with these catalysts. Complexes of aluminum chloride and methylaluminum dichloride with bis-(cyclopentadienyl)-titanium dichloride as catalysts for the polymerization of ethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
One-pot synthesis of α-methylene-γ-butyrolactones from benzaldehydes and bromomethylacrylates. Bis-(cyclopentadienyl)-titanium dichloride alkylaluminum complexls acts as catalysts for the polymerization of ethylene. Polyethylene prepared with these catalysts. Complexes of aluminum chloride and methylaluminum dichloride with bis-(cyclopentadienyl)-titanium dichloride as catalysts for the polymerization of ethylene.

Solubility
Slightly soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, air.
RUO – Research Use Only

General References:

  1. Wendell P. Long,; David S. Breslow. Polymerization of Ethylene with Bis-(cyclopentadienyl)-titanium Dichloride and Diethylaluminum Chloride. J. Am. Chem. Soc. 1960, 82 (8), 1953-1957.
  2. Wendell P. Long. Complexes of Aluminum Chloride and Methylaluminum Dichloride with Bis-(cyclopentadienyl)-titanium Dichloride as Catalysts for the Polymerization of Ethylene . J. Am. Chem. Soc. 1959, 81 (20),5312-5316.
  3. In combination with trimethyaluminum, generates an ylide (Tebbe reagent): J. Am. Chem. Soc., 100, 3611 (1978), which transfers a methylene group not only to aldehydes and ketones (cf Wittig reaction) but also to the carbonyl groups of esters and lactones: J. Am. Chem. Soc., 102, 3270 (1980):
  4. A similar reagent can be prepared by reaction with the Simmons-Smith adduct (see Diiodomethane, A15457) and used for the methylenation of ketones in excellent yields: Tetrahedron Lett., 24, 2043 (1983).
  5. Reacts with Grignard reagents with a ß-hydrogen atom (e.g. isobutyl or isopropyl) to form, in a catalytic cycle, a titanium hydride species, which reduces aldehydes, ketones and esters to alcohols: Tetrahedron Lett., 21, 2171, 2175 (1980). The same combination effects regioselective cis-hydromagnesation of alkynes to give alkenyl Grignard reagents: Tetrahedron Lett., 22, 85 (1981); J. Chem. Soc., Chem. Commun., 718 (1981). For application in a highly stereospecific synthesis of vinylsilanes in excellent yields, see: Tetrahedron Lett., 24, 1041 (1983):
  6. For further examples of the hydroxymagnesation of alkynes, see: Org. Synth. Coll., 8, 507 (1993). Similarly π-allyl complexes with 1,3-dienes can be generated, which react regio-and stereo-selectively with aldehydes to give homoallylic alcohols: J. Chem. Soc., Chem. Commun., 621 (1984):
  7. For use, in combination with Mg metal, in formation of titanocycle intermediates, see: J. Org. Chem., 63, 9285 (1998); for reaction scheme, see Triphosgene, A14932.